Learn More
Cayman Chemical (S)-(−)-5-Fluorowillardiine (hydrochloride), Purity: ≥95%, CAS: 1321546-70-6, 10 mg
SDP
Catalog No. 502919328
Encompass_Preferred
An agonist of GluA1 and GluA2; selectively activates GluA1 and GluA2 over GluA3 and GluA4 in voltage-clamp assays using X. laevis oocytes expressing the flip isoforms of the rat receptors (EC50s = 0.382, 0.463, 20.9, and 11.9 µM, respectively); induces evoked population spikes in isolated rat CA1 hippocampal slices in an in vitro model of seizure-like activity (EC50 = 0.03-0.08 µM)
Catalog No. 50-291-9328 Supplier Cayman Chemical Supplier No. 3737210 MG
An agonist of GluA1 and GluA2; selectively activates GluA1 and GluA2 over GluA3 and GluA4 in voltage-clamp assays using X. laevis oocytes expressing the flip isoforms of the rat receptors (EC50s = 0.382, 0.463, 20.9, and 11.9 µM, respectively); induces evoked population spikes in isolated rat CA1 hippocampal slices in an in vitro model of seizure-like activity (EC50 = 0.03-0.08 µM)