Learn More
Custom Biogenic Systems S-5000ABEH LN2 Freezer, 720 Liters Capacity, 46,500 Vials, Castor-Mounted
Catalog No. NC3652587
Encompass
The Custom Biogenic Systems S-5000ABEH is a high-capacity LN2 freezer designed for the secure cryogenic storage of biological samples. Featuring a robust stainless steel design and integrated autofill-monitor system, it ensures reliable operation and sample protection. With a maximum capacity of 46,500 vials and 720 liters of LN2, it is ideal for biobanking and long-term preservation. Its mobility and space-saving design differentiate it from traditional models, offering practical benefits for laboratory environments.
Catalog No. NC3652587 Supplier Custom Biogenic Systems Supplier No. 33505BEH
Add to Cart
Add to Cart
Request a Quote
The Custom Biogenic Systems S-5000ABEH is a high-capacity LN2 freezer designed for the secure cryogenic storage of biological samples. Featuring a robust stainless steel design and integrated autofill-monitor system, it ensures reliable operation and sample protection. With a maximum capacity of 46,500 vials and 720 liters of LN2, it is ideal for biobanking and long-term preservation. Its mobility and space-saving design differentiate it from traditional models, offering practical benefits for laboratory environments.