Learn More
Cayman Chemical S-(5'-Adenosyl)-L-methionine chloride (hydrochloride), Purity: ≥95%, CAS: 86867-01-8, 2.5 mg
SDP
Catalog No. 502954475
Encompass_Preferred
A ubiquitous methyl donor involved in a wide variety of biological reactions, including those mediated by DNA and protein methyltransferases
Catalog No. 50-295-4475 Supplier Cayman Chemical Supplier No. 139562.5 MG
A ubiquitous methyl donor involved in a wide variety of biological reactions, including those mediated by DNA and protein methyltransferases