Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-6-Oxo-2-Piperidinecarboxylic Acid

Catalog No. 363235GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-323-5GF 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 36-323-5GF Supplier Sigma Aldrich Supplier No. 363235GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-6-Oxo-2-piperidinecarboxylic acid can be used as a reactant to synthesize: Functionalized +a-lactam N-heterocycles via carboxymethylproline synthase catalyzed cyclization reactions. Pro-(S)-C5 branched [4.3.1] aza-amide bicycles as potential inhibitors for FK506-binding proteins. Ethyl (S)-2-(6-oxopiperidin-2-yl)acetate, which is reduced using LiBH4 to produce optically pure hydroxymethyl lactams.

Specifications

CAS 66943-05-3
Melting Point 35°C to 37°C
Molecular Formula C10H21NO4
Linear Formula C10H21NO4
MDL Number MFCD00075465
Quantity 1 g
Synonym 6-Oxo-L-pipecolic acid; (S)-2-Piperidone-6-carboxylic acid; L-Pyrohomoglutamic acid
Molecular Weight (g/mol) 219.28 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.