Learn More
Grainger Hood with Shoulder Coverage: Versaflo, Hood, PAPR System/SAR System, Universal Headgear Size, 3M Mfr. Model S-655
Catalog No. NC3717388
Encompass_Preferred
3M general purpose PAPR (powered air-purifying respirator) headgear covers the head to allow clean air to be directed to the worker. A breathing tube connects the helmet to the filtered air source to deliver a constant flow of air inside the worker's headgear.
Catalog No. NC3717388 Supplier Grainger Supplier No. 5WZD0
3M general purpose PAPR (powered air-purifying respirator) headgear covers the head to allow clean air to be directed to the worker. A breathing tube connects the helmet to the filtered air source to deliver a constant flow of air inside the worker's headgear.