Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-7-ethyl-7-hydroxy-14-(3-hydroxypropyl)-10,13-dihydro-11H-dioxolo[4,5-g]pyrano[3',4':6,7]i... | 2821768-98-1 | 98.7% | 450.44 g/mol | C24H22N2O7 | 1 ML
SDP

Catalog No. 5000204465
Encompass_Preferred

FL118-14-Propanol is a synthetic derivative of FL118 provided as a research reagent. It is supplied as a 10 mM solution in DMSO and has reported anti-tumor activity in mouse models. Intended for research use only.

  • IUPAC name: (S)-7-ethyl-7-hydroxy-14-(3-hydroxypropyl)-10,13-dihydro-11H-dioxolo[4,5-g]pyrano[3',4':6,7]indolizino[1,2-b]quinoline-8,11(7H)-dione.
  • Cas number: 2821768-98-1.
  • Chemical formula: C24H22N2O7.
  • Molecular weight: 450.44 g/mol.
  • Purity: 98.69% (reported by manufacturer).
  • Formulation: 10 mM solution in DMSO, 1 mL vial.
  • Solubility: DMSO up to 100 mg/mL (≈222.01 mM); ultrasonic assistance may be required.
  • Storage: protect from light; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-04465 Supplier Medchemexpress LLC Supplier No. HY15685610MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FL118-14-Propanol is a synthetic derivative of FL118 provided as a research reagent. It is supplied as a 10 mM solution in DMSO and has reported anti-tumor activity in mouse models. Intended for research use only.

  • IUPAC name: (S)-7-ethyl-7-hydroxy-14-(3-hydroxypropyl)-10,13-dihydro-11H-dioxolo[4,5-g]pyrano[3',4':6,7]indolizino[1,2-b]quinoline-8,11(7H)-dione.
  • Cas number: 2821768-98-1.
  • Chemical formula: C24H22N2O7.
  • Molecular weight: 450.44 g/mol.
  • Purity: 98.69% (reported by manufacturer).
  • Formulation: 10 mM solution in DMSO, 1 mL vial.
  • Solubility: DMSO up to 100 mg/mL (≈222.01 mM); ultrasonic assistance may be required.
  • Storage: protect from light; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.