Learn More
Medchemexpress LLC (S)-9b-(4-chloro-3-methylphenyl)-1-(3,4-difluorobenzoyl)-1,2,3,9b-tetrahydro-5H-imidazo[2,1-a]isoind | 2092923-21-0 | 99.5% | 438.85 | C24H17ClF2N2O2 | 10 MM 1 ML
SDP
Catalog No. 5000211664
Encompass_Preferred
VU 6008667 is a selective negative allosteric modulator of the muscarinic acetylcholine receptor M5. It inhibits human M5 and rat M5 with IC50 values of 1.2 μM and 1.6 μM, respectively, and is supplied for research use only as a 10 mM solution in DMSO (1 mL); solid quantities are also available. Reported data indicate high central nervous system penetration.
  • Selective M5 negative allosteric modulator with documented IC50 values.
  • High central nervous system penetration suitable for CNS studies.
  • Provided as a ready-to-use 10 mM solution in DMSO (1 mL).
  • High purity with supporting analytical documentation (COA, HPLC, MS).
  • Intended for research use only; not for clinical or therapeutic use.
Catalog No. 50-002-11664 Supplier Medchemexpress LLC Supplier No. HY10128110MM 1ML
VU 6008667 is a selective negative allosteric modulator of the muscarinic acetylcholine receptor M5. It inhibits human M5 and rat M5 with IC50 values of 1.2 μM and 1.6 μM, respectively, and is supplied for research use only as a 10 mM solution in DMSO (1 mL); solid quantities are also available. Reported data indicate high central nervous system penetration.
  • Selective M5 negative allosteric modulator with documented IC50 values.
  • High central nervous system penetration suitable for CNS studies.
  • Provided as a ready-to-use 10 mM solution in DMSO (1 mL).
  • High purity with supporting analytical documentation (COA, HPLC, MS).
  • Intended for research use only; not for clinical or therapeutic use.