Learn More
Cayman Chemical (S)-α-Methylbenzyl Ricinoleamide, Purity: ≥98%, CAS: 1246776-22-6, 10 mg
SDP
Catalog No. 502966103
Encompass_Preferred
A fatty acid amide derived from ricinoleic acid and methyl benzylamine that demonstrates potent growth inhibition of glioma (U251), breast (MCF-7), ovarian (NCI-ADR/RES and OVCAR-3), kidney (786-0), non-small cell lung (NCI-H460), and prostate (PC-3) cancer cells with a mean GI50 value of 6.9 µM
Catalog No. 50-296-6103 Supplier Cayman Chemical Supplier No. 1754610 MG
A fatty acid amide derived from ricinoleic acid and methyl benzylamine that demonstrates potent growth inhibition of glioma (U251), breast (MCF-7), ovarian (NCI-ADR/RES and OVCAR-3), kidney (786-0), non-small cell lung (NCI-H460), and prostate (PC-3) cancer cells with a mean GI50 value of 6.9 µM