Learn More
Medchemexpress LLC (S)-ACE-OH | 99.2% | 328.47 | 50 MG
SDP
Catalog No. 5000390849
Encompass_Preferred
(S)-ACE-OH is identified as a molecular glue that exhibits anticancer activity. Its mechanism of action involves promoting the degradation of nucleoporin and disrupting nucleocytoplasmic trafficking. This is achieved by inducing an interaction between the E3 ubiquitin ligase TRIM21 and nucleoporin NUP98. It is intended for research use only.
  • Acts as a molecular glue with anticancer properties.
  • Promotes nucleoporin degradation.
  • Disrupts nucleocytoplasmic trafficking.
  • Induces interaction between E3 ubiquitin ligase TRIM21 and nucleoporin NUP98.
Catalog No. 50-003-90849 Supplier Medchemexpress LLC Supplier No. HY16932550MG
(S)-ACE-OH is identified as a molecular glue that exhibits anticancer activity. Its mechanism of action involves promoting the degradation of nucleoporin and disrupting nucleocytoplasmic trafficking. This is achieved by inducing an interaction between the E3 ubiquitin ligase TRIM21 and nucleoporin NUP98. It is intended for research use only.
  • Acts as a molecular glue with anticancer properties.
  • Promotes nucleoporin degradation.
  • Disrupts nucleocytoplasmic trafficking.
  • Induces interaction between E3 ubiquitin ligase TRIM21 and nucleoporin NUP98.