Learn More
Cayman Chemical S-Acenocoumarol 10mg
SDP
Catalog No. 502894011
Encompass_Preferred
An enantiomer of the short-lived oral anti-coagulant acenocoumarol with a shorter plasma elimination half-life (1.8 hours) and faster plasma clearance (28.5 L/hour), compared to the (R)-enantiomer (6.6 hours, 1.9 L/hour); undergoes extensive first-pass metabolism during absorption from the gastrointestinal tract; less potent in vivo than the (R)-enantiomer
Catalog No. 50-289-4011 Supplier Cayman Chemical Supplier No. 900033610 MG
An enantiomer of the short-lived oral anti-coagulant acenocoumarol with a shorter plasma elimination half-life (1.8 hours) and faster plasma clearance (28.5 L/hour), compared to the (R)-enantiomer (6.6 hours, 1.9 L/hour); undergoes extensive first-pass metabolism during absorption from the gastrointestinal tract; less potent in vivo than the (R)-enantiomer