Learn More
Medchemexpress LLC S-acetyl-dPEG4-NHS ester | 937025-17-7 | MFCD11041102 | 98.0% | 421.46 g/mol | C17H27NO9S | 10 MG
SDP
Catalog No. 5000281590
Encompass_Preferred
S-acetyl-dPEG4-NHS ester is a discrete polyethylene glycol reagent that installs a protected thiol group via an N-hydroxysuccinimide (NHS) ester. It is used to thiolate proteins, peptides, and small molecules to enable downstream conjugation and crosslinking in bioconjugation and drug-delivery research. The reagent is supplied as a solid with reported purity and molecular weight.
  • Protected thiol for controlled thiolation reactions.
  • NHS-ester reactive group for efficient coupling to amines.
  • Discrete PEG4 spacer improves solubility and reduces steric hindrance.
  • Facilitates protein, peptide, and small-molecule conjugation.
  • Suitable for bioconjugation and drug-delivery research workflows.
  • Supplied as a stable solid suitable for storage and handling.
Catalog No. 50-002-81590 Supplier Medchemexpress LLC Supplier No. HYW19074010MG
S-acetyl-dPEG4-NHS ester is a discrete polyethylene glycol reagent that installs a protected thiol group via an N-hydroxysuccinimide (NHS) ester. It is used to thiolate proteins, peptides, and small molecules to enable downstream conjugation and crosslinking in bioconjugation and drug-delivery research. The reagent is supplied as a solid with reported purity and molecular weight.
  • Protected thiol for controlled thiolation reactions.
  • NHS-ester reactive group for efficient coupling to amines.
  • Discrete PEG4 spacer improves solubility and reduces steric hindrance.
  • Facilitates protein, peptide, and small-molecule conjugation.
  • Suitable for bioconjugation and drug-delivery research workflows.
  • Supplied as a stable solid suitable for storage and handling.