Learn More
Medchemexpress LLC S-acetyl-PEG3-alcohol | 153870-20-3 | MFCD22574787 | >98.0% | 208.28 g/mol | C8H16O4S | 5 MG
SDP
Catalog No. 5000223208
Encompass_Preferred
S-acetyl-PEG3-alcohol is a short polyethylene glycol (PEG3) linker bearing an S-acetyl protected thiol and a terminal hydroxyl group. It is provided for research use and is commonly used as a spacer in the chemical synthesis of PROTAC molecules. Molecular formula C8H16O4S; molecular weight 208.28 g/mol.
  • Provides a PEG3 spacer for linker construction.
  • S-acetyl protects the thiol for controlled deprotection and conjugation.
  • Terminal alcohol enables further functionalization or coupling reactions.
  • Available in small research quantities suitable for medicinal chemistry.
  • Suitable for PROTAC synthesis and linker optimization.
Catalog No. 50-002-23208 Supplier Medchemexpress LLC Supplier No. HY1413405MG
S-acetyl-PEG3-alcohol is a short polyethylene glycol (PEG3) linker bearing an S-acetyl protected thiol and a terminal hydroxyl group. It is provided for research use and is commonly used as a spacer in the chemical synthesis of PROTAC molecules. Molecular formula C8H16O4S; molecular weight 208.28 g/mol.
  • Provides a PEG3 spacer for linker construction.
  • S-acetyl protects the thiol for controlled deprotection and conjugation.
  • Terminal alcohol enables further functionalization or coupling reactions.
  • Available in small research quantities suitable for medicinal chemistry.
  • Suitable for PROTAC synthesis and linker optimization.