Learn More
Medchemexpress LLC S-acetyl-PEG4-amine (TFA) | 00-00-0 | 98.0% | 409.42 g/mol | C14H26F3NO7S | 5 MG
SDP
Catalog No. 5000222916
Encompass_Preferred
S-acetyl-PEG4-amine (TFA) is a PEG-based PROTAC linker used in the chemical synthesis of proteolysis-targeting chimeras (PROTACs). PROTACs are heterobifunctional molecules that recruit an E3 ubiquitin ligase to a target protein, promoting ubiquitination and proteasomal degradation of the target.
  • Provides a short PEG4 spacer to control linker length and flexibility.
  • Contains a terminal amine (supplied as the TFA salt) for amide coupling and other conjugation chemistries.
  • Features an S-acetyl protecting group to allow selective deprotection strategies.
  • Available in small-quantity packages suitable for synthesis optimization and screening.
Catalog No. 50-002-22916 Supplier Medchemexpress LLC Supplier No. HY138752A5MG
S-acetyl-PEG4-amine (TFA) is a PEG-based PROTAC linker used in the chemical synthesis of proteolysis-targeting chimeras (PROTACs). PROTACs are heterobifunctional molecules that recruit an E3 ubiquitin ligase to a target protein, promoting ubiquitination and proteasomal degradation of the target.
  • Provides a short PEG4 spacer to control linker length and flexibility.
  • Contains a terminal amine (supplied as the TFA salt) for amide coupling and other conjugation chemistries.
  • Features an S-acetyl protecting group to allow selective deprotection strategies.
  • Available in small-quantity packages suitable for synthesis optimization and screening.