Learn More
Medchemexpress LLC S-acetyl-PEG4-propargyl | 1422540-88-2 | 97.0% | C13H22O5S | 250 MG
SDP
Catalog No. 5000357359
Encompass_Preferred
S-acetyl-PEG4-propargyl is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are compounds with two ligands connected by a linker: one targets an E3 ubiquitin ligase, and the other targets a protein of interest. This design enables PROTACs to utilize the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • Functions as a PEG-based PROTAC linker.
  • Enables the synthesis of PROTAC compounds.
  • Supports targeted protein degradation.
  • Utilizes the ubiquitin-proteasome system.
Catalog No. 50-003-57359 Supplier Medchemexpress LLC Supplier No. 250MGHY141350
S-acetyl-PEG4-propargyl is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are compounds with two ligands connected by a linker: one targets an E3 ubiquitin ligase, and the other targets a protein of interest. This design enables PROTACs to utilize the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • Functions as a PEG-based PROTAC linker.
  • Enables the synthesis of PROTAC compounds.
  • Supports targeted protein degradation.
  • Utilizes the ubiquitin-proteasome system.