Learn More
Sigma Aldrich S-Acetylthioglycolic acid N-hydroxysuccinimide ester
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Thiolating reagent for primary amines; thiol can be deprotected under mild conditions
- Typically, couples initially to a molecule containing primary amine by amide bond buffered at pH 7.5
- Deprotection may be accomplished with 0.05 m hydroxylamine at neutral pH
- Useful for preparation of enzyme immunoconjugates and hapten carrier molecule conjugates.note that the ester couples very efficiently to primary amines compared to other thiolating reagents
- In addition, this reagent will neutralize original amine positive charge
Specifications
Specifications
| CAS | 76931-93-6 |
| Molecular Formula | C8H9NO5S |
| Linear Formula | C8H9NO5S |
| MDL Number | MFCD00036891 |
| Quantity | 100 mg |
| Synonym | N-Succinimidyl (acetylthio)acetate; N-Succinimidyl S-acetylthioglycolate; S-Acetylthioglycolic acid NHS ester; SATA |
| Molecular Weight (g/mol) | 231.23 |
| Percent Purity | ≥95% (TLC) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.