Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich S-Acetylthioglycolic acid N-hydroxysuccinimide ester

Catalog No. A904350MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A904350MG 50 mg
A904310MG 10 mg
A9043100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A904350MG Supplier Sigma Aldrich Supplier No. A904350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Thiolating reagent for primary amines; thiol can be deprotected under mild conditions
  • Typically, couples initially to a molecule containing primary amine by amide bond buffered at pH 7.5
  • Deprotection may be accomplished with 0.05 m hydroxylamine at neutral pH
  • Useful for preparation of enzyme immunoconjugates and hapten carrier molecule conjugates.note that the ester couples very efficiently to primary amines compared to other thiolating reagents
  • In addition, this reagent will neutralize original amine positive charge

Specifications

CAS 76931-93-6
Molecular Formula C8H9NO5S
Linear Formula C8H9NO5S
MDL Number MFCD00036891
Quantity 50 mg
Synonym N-Succinimidyl (acetylthio)acetate; N-Succinimidyl S-acetylthioglycolate; S-Acetylthioglycolic acid NHS ester; SATA
Molecular Weight (g/mol) 231.23
Percent Purity ≥95% (TLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.