Learn More
Raybiotech Inc RAYBIOTECH INC S-Acylation Detection Kit, Biotin Switch, 40 Sample Kit
Catalog No. NC3997721
Encompass_Preferred
The S-Acylation Detection Kit employs a modified biotin-switch method to enable direct visualization of S-acylation, including S-palmitoylation, of proteins via western blot analysis. This kit is capable of biotin-labeling 40 samples, each containing 100-200 μg of total protein.
  • Utilizes modified biotin-switch method for S-acylation visualization.
  • Detects S-palmitoylation of proteins via western blot analysis.
  • Biotin-labels 40 samples, each with 100 to 200 μg of protein.
  • Employs acyl-biotinyl exchange (ABE) method.
  • Blocks unmodified free cysteines, then selectively labels acylated cysteines.
  • Biotinylation detected by western blot or immunoassay.
  • Avidin resin enriches biotin-labeled acylated proteins/peptides.
  • Facilitates S-acylation/palmitoylation site mapping via mass spectrometry.
Catalog No. NC3997721 Supplier Raybiotech Inc Supplier No. MAACYLW
The S-Acylation Detection Kit employs a modified biotin-switch method to enable direct visualization of S-acylation, including S-palmitoylation, of proteins via western blot analysis. This kit is capable of biotin-labeling 40 samples, each containing 100-200 μg of total protein.
  • Utilizes modified biotin-switch method for S-acylation visualization.
  • Detects S-palmitoylation of proteins via western blot analysis.
  • Biotin-labels 40 samples, each with 100 to 200 μg of protein.
  • Employs acyl-biotinyl exchange (ABE) method.
  • Blocks unmodified free cysteines, then selectively labels acylated cysteines.
  • Biotinylation detected by western blot or immunoassay.
  • Avidin resin enriches biotin-labeled acylated proteins/peptides.
  • Facilitates S-acylation/palmitoylation site mapping via mass spectrometry.