Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC S-Adenosyl-L-methionine-13C dihydrochloride | 29908-03-0 | 98.64% | 20 MG
SDP

Catalog No. 5000408312
Encompass_Preferred

S-Adenosyl-L-methionine-13C dihydrochloride is the 13C labeled version of S-Adenosyl-L-methionine, an orally active methyl group donor and dietary supplement. It has potent antidepressant effects and exhibits anti-proliferative, pro-apoptotic, and anti-metastatic roles in cancers, with potential for research in cancer, liver disease, and osteoarthritis.

  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Molecular Weight: 508.81
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Solubility: In H2O: 50 mg/mL (98.27 mM; requires ultrasonic assistance)

Catalog No. 50-004-08312 Supplier Medchemexpress LLC Supplier No. HYB0617S220MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S-Adenosyl-L-methionine-13C dihydrochloride is the 13C labeled version of S-Adenosyl-L-methionine, an orally active methyl group donor and dietary supplement. It has potent antidepressant effects and exhibits anti-proliferative, pro-apoptotic, and anti-metastatic roles in cancers, with potential for research in cancer, liver disease, and osteoarthritis.

  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Molecular Weight: 508.81
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Solubility: In H2O: 50 mg/mL (98.27 mM; requires ultrasonic assistance)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.