Learn More
Medchemexpress LLC S-Adenosyl-L-methionine | 29908-03-0 | 99.7% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB061750MG
Encompass_Preferred
S-Adenosyl-L-methionine (S-Adenosyl methionine) is an orally active methyl group donor. It functions as a dietary supplement with potent antidepressant effects. Furthermore, it demonstrates anti-proliferative, pro-apoptotic, and anti-metastatic activities in cancers, and shows potential for research in cancer, liver disease, and osteoarthritis.
  • Orally active methyl group donor.
  • Dietary supplement with potent antidepressant effects.
  • Exhibits anti-proliferative, pro-apoptotic, and anti-metastatic roles in cancers.
  • Potential for research in cancer.
  • Potential for research in liver disease.
  • Potential for research in osteoarthritis.
Catalog No. 50-004-08320