Learn More
Medchemexpress LLC S-Adenosyl-L-methionine | 29908-03-0 | 99.96% | 5 MG
SDP
Catalog No. 5000405911
Encompass_Preferred
S-Adenosyl-L-methionine is an orally active methyl group donor and dietary supplement with potent antidepressant effects. It plays anti-proliferative, pro-apoptotic, and anti-metastatic roles in cancers. This compound has potential for research in cancer, liver disease, and osteoarthritis.
  • Induces cell apoptosis and promotes cell cycle arrest
  • Decreases cell migration
  • Protects anticancer effects of 5-FU
  • Prevents ASD-like behaviors in mice
  • Shows antiepileptic, memory-enhancing, and antioxidant properties
Catalog No. 50-004-05911 Supplier Medchemexpress LLC Supplier No. HYB06175MG
S-Adenosyl-L-methionine is an orally active methyl group donor and dietary supplement with potent antidepressant effects. It plays anti-proliferative, pro-apoptotic, and anti-metastatic roles in cancers. This compound has potential for research in cancer, liver disease, and osteoarthritis.
  • Induces cell apoptosis and promotes cell cycle arrest
  • Decreases cell migration
  • Protects anticancer effects of 5-FU
  • Prevents ASD-like behaviors in mice
  • Shows antiepileptic, memory-enhancing, and antioxidant properties