Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC S-adenosyl-L-methionine | 29908-03-0 | MFCD00871208 | 10 MG
SDP

Catalog No. 5000297369
Encompass_Preferred

S-adenosyl-L-methionine (SAMe) is an endogenous methyl group donor used in biochemical and pharmacological research. The compound is supplied for research and analytical applications and is provided in small milligram pack sizes suitable for assay development and method validation.

  • Endogenous methyl group donor used in methylation studies.
  • Molecular formula C15H22N6O5S; molecular weight 398.44 g/mol.
  • Applicable to biochemical, pharmacological, and liver disease research.
  • Available in small-mass pack sizes for assay development.
  • Manufacturer datasheet available for specifications and safe handling.

Catalog No. 50-002-97369 Supplier Medchemexpress LLC Supplier No. HYB0617S210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S-adenosyl-L-methionine (SAMe) is an endogenous methyl group donor used in biochemical and pharmacological research. The compound is supplied for research and analytical applications and is provided in small milligram pack sizes suitable for assay development and method validation.

  • Endogenous methyl group donor used in methylation studies.
  • Molecular formula C15H22N6O5S; molecular weight 398.44 g/mol.
  • Applicable to biochemical, pharmacological, and liver disease research.
  • Available in small-mass pack sizes for assay development.
  • Manufacturer datasheet available for specifications and safe handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.