Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC S-adenosyl-L-methionine | 29908-03-0 | MFCD00871208 | ≥98.0% | 398.44 g/mol | C15H22N6O5S | 10 MG
SDP

Catalog No. 5000298115
Encompass_Preferred

S-Adenosyl-L-methionine (SAMe) is an endogenous methyl group donor used in biochemical research and as a dietary supplement. This high-purity reagent is supplied as an off-white to light yellow solid suitable for methylation studies, enzymology, and biological assays.

  • Purity ≥98.0%.
  • Molecular weight 398.44 g/mol.
  • Chemical formula C15H22N6O5S.
  • Appearance off-white to light yellow solid.
  • Recommended storage -20°C; protect from light.
  • Common applications: methyl group donor for methylation reactions, and research into antidepressant effects, liver disease, and osteoarthritis.

Catalog No. 50-002-98115 Supplier Medchemexpress LLC Supplier No. HYB061710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S-Adenosyl-L-methionine (SAMe) is an endogenous methyl group donor used in biochemical research and as a dietary supplement. This high-purity reagent is supplied as an off-white to light yellow solid suitable for methylation studies, enzymology, and biological assays.

  • Purity ≥98.0%.
  • Molecular weight 398.44 g/mol.
  • Chemical formula C15H22N6O5S.
  • Appearance off-white to light yellow solid.
  • Recommended storage -20°C; protect from light.
  • Common applications: methyl group donor for methylation reactions, and research into antidepressant effects, liver disease, and osteoarthritis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.