Learn More
Medchemexpress LLC S-Adenosyl-L-methionine chloride dihydrochloride | 86867-01-8 | 75.0% | 507.82 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14614A50MG
Encompass_Preferred
S-Adenosyl-L-methionine chloride dihydrochloride is an orally active methyl group donor. It is a dietary supplement with potent antidepressant effects and also demonstrates anti-proliferative, pro-apoptotic, and anti-metastatic roles in cancers. This compound has potential for research in cancer, liver disease, and osteoarthritis.
  • Induces cell apoptosis and promotes cell cycle arrest
  • Decreases migration of cells
  • Protects anticancer effect of 5-FU
  • Prevents ASD-like behaviors
  • Shows antiepileptic, memory-enhancing, and antioxidant properties
Catalog No. 50-003-60292