Learn More
Medchemexpress LLC S-adenosyl-L-methionine chloride dihydrochloride | 86867-01-8 | FCD22571809 | 75.0% | 507.82 g/mol | C15H25Cl3N6O5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY14614A5MG
Encompass_Preferred
S-Adenosyl-L-methionine chloride dihydrochloride is the dihydrochloride salt of SAMe, a biological methyl donor used in biochemical and pharmacological research. Supplied as a white to light yellow solid, it is used to study methylation reactions, neurotransmitter metabolism, and related enzymatic pathways. Store sealed at -20 °C and protect from moisture; in solution, store at -80 °C up to 6 months or -20 °C up to 1 month.
  • Provides an active methyl donor for biochemical assays.
  • Useful for studies of methylation and neurotransmitter metabolism.
  • White to light yellow solid for straightforward handling and weighing.
  • Reported purity of 75.0% for experimental reproducibility.
  • Recommended frozen storage to preserve stability and activity.
Catalog No. 50-002-22822