Learn More
Medchemexpress LLC S-adenosyl-L-methionine chloride dihydrochloride | 86867-01-8 | FCD22571809 | 75.0% | C15H25Cl3N6O5S | 10MG
SDP
Catalog No. 5000258729
Encompass_Preferred
S-adenosyl-L-methionine chloride dihydrochloride is the chloride dihydrochloride salt of S-adenosyl-L-methionine (SAM), a biological methyl donor commonly used in biochemical and pharmacological research. Supplied as a laboratory reagent in small package sizes, it is employed in enzymatic methylation assays, supplementation studies, and metabolic investigations.
  • Acts as a methyl group donor in enzymatic methylation reactions.
  • Suitable for biochemical and pharmacological research and in vitro assays.
  • Available in small laboratory package sizes for precise dosing.
  • Often used in studies of neurotransmitter metabolism and liver function.
  • Typical supplied purity is 75.0%.
Catalog No. 50-002-58729 Supplier Medchemexpress LLC Supplier No. HY14614A10MG
S-adenosyl-L-methionine chloride dihydrochloride is the chloride dihydrochloride salt of S-adenosyl-L-methionine (SAM), a biological methyl donor commonly used in biochemical and pharmacological research. Supplied as a laboratory reagent in small package sizes, it is employed in enzymatic methylation assays, supplementation studies, and metabolic investigations.
  • Acts as a methyl group donor in enzymatic methylation reactions.
  • Suitable for biochemical and pharmacological research and in vitro assays.
  • Available in small laboratory package sizes for precise dosing.
  • Often used in studies of neurotransmitter metabolism and liver function.
  • Typical supplied purity is 75.0%.