Learn More
Medchemexpress LLC S-adenosyl-L-methionine disulfate tosylate | 97540-22-2 | MFCD01674068 | 99.5% | 766.80 g/mol | C22H34N6O16S4 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW017770
Encompass_Preferred
S-adenosyl-L-methionine disulfate tosylate is the disulfate tosylate salt form of S-adenosyl-L-methionine, a biologically active methyl group donor used in biochemical and pharmacological research. It is provided as a high-purity solid for use in studies of methylation, neurotransmitter synthesis, liver function, and cancer-related pathways.
  • High purity: 99.5%.
  • Solid, white to light yellow form for easy handling and storage.
  • Functionally active methyl donor for epigenetic and metabolic assays.
  • Storage-stable when sealed at low temperatures; guidelines provided for solvent storage.
  • Suitable for in vitro and in vivo research applications.
  • Available in small research quantities for laboratory use.
Catalog No. 50-003-02491