Learn More
Medchemexpress LLC S-Adenosyl-L-methionine tosylate | 52248-03-0 | 99.6% | 1 ML
SDP
Catalog No. 5000405813
Encompass_Preferred
S-Adenosyl-L-methionine tosylate is an orally active methyl group donor with potent antidepressant effects. It demonstrates anti-proliferative, pro-apoptotic, and anti-metastatic roles in cancers, and has potential for research in cancer, liver disease, and osteoarthritis.
  • Orally active methyl group donor
  • Potent antidepressant effects
  • Anti-proliferative, pro-apoptotic, and anti-metastatic roles in cancers
  • Potential for research in cancer, liver disease, and osteoarthritis
  • Induces cell apoptosis and promotes cell cycle arrest in vitro
  • Decreases migration of cancer cells in vitro
  • Protects anticancer effect of 5-FU in vitro
  • Alleviates ASD-like neurobehavioral symptoms in animal models
  • Shows antiepileptic, memory-enhancing, and antioxidant properties in animal models
Catalog No. 50-004-05813 Supplier Medchemexpress LLC Supplier No. HYB0617A10MM/1ML
S-Adenosyl-L-methionine tosylate is an orally active methyl group donor with potent antidepressant effects. It demonstrates anti-proliferative, pro-apoptotic, and anti-metastatic roles in cancers, and has potential for research in cancer, liver disease, and osteoarthritis.
  • Orally active methyl group donor
  • Potent antidepressant effects
  • Anti-proliferative, pro-apoptotic, and anti-metastatic roles in cancers
  • Potential for research in cancer, liver disease, and osteoarthritis
  • Induces cell apoptosis and promotes cell cycle arrest in vitro
  • Decreases migration of cancer cells in vitro
  • Protects anticancer effect of 5-FU in vitro
  • Alleviates ASD-like neurobehavioral symptoms in animal models
  • Shows antiepileptic, memory-enhancing, and antioxidant properties in animal models