Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC S-Adenosyl-L-methionine (tosylate) | 52248-03-0 | 99.6% | 50 MG
SDP

Supplier:  Medchemexpress LLC HYB0617AR50MG

Encompass_Preferred

S-Adenosyl-L-methionine tosylate is an orally active methyl group donor and dietary supplement with potent antidepressant effects. This compound also exhibits anti-proliferative, pro-apoptotic, and anti-metastatic roles in various cancers. It has potential for research in cancer, liver disease, and osteoarthritis, and is utilized in laboratory chemical applications and substance manufacturing.

  • Orally active methyl group donor.
  • Dietary supplement with potent antidepressant effects.
  • Exhibits anti-proliferative, pro-apoptotic, and anti-metastatic roles in various cancers.
  • Potential for research in cancer, liver disease, and osteoarthritis.
  • Suitable for laboratory chemical applications and substance manufacturing.
  • Appearance: Solid.
  • Molecular weight: 570.64.

Catalog No. 50-004-05912


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.