Learn More
Cayman Chemical S-Adenosylhomocysteine-d4, Purity: ≥99% deuterated forms (d1-d4), 1 mg
SDP
Catalog No. 502896558
Encompass_Preferred
An internal standard for the quantification of SAH by GC- or LC-MS
Catalog No. 50-289-6558 Supplier Cayman Chemical Supplier No. 90003721 MG
An internal standard for the quantification of SAH by GC- or LC-MS