Learn More
Cayman Chemical S-Adenosylhomocysteine-d4, Purity: ≥99% deuterated forms (d1-d4), 5 mg
SDP
Catalog No. 502897005
Encompass_Preferred
An internal standard for the quantification of SAH by GC- or LC-MS
Catalog No. 50-289-7005 Supplier Cayman Chemical Supplier No. 90003725 MG
An internal standard for the quantification of SAH by GC- or LC-MS