Learn More
Cayman Chemical S-Adenosylhomocysteine, 979-92-0, 10 mg
SDP
Supplier:  Cayman Chemical 1360310
Encompass_Preferred
Molecular Formula C14H20N6O5S, Purity ≥98%, Formula Weight 384.4, Synonyms: AdoHcy, SAH; An amino acid derivative and an intermediate, by-product, or modulator of several metabolic pathways, including the activated methyl cycle and cysteine biosynthesis; also a product of SAM-dependent methylation of biological molecules, including DNA, RNA, and histones, and other proteins.
Catalog No. NC0503925