Learn More
New England Biolabs, Inc. S-adenosylmethionine (SAM) – 0.5 ml
SDP
Catalog No. 50811564
Encompass
S-adenosyl-methionine (SAM) is included with all of the DNA methylases available from New England Biolabs, Inc. as well as certain restriction enzymes for which it is required. The SAM is prepared in a solution of 0.005 M H2SO4 and 10% ETOH and filtered.
This item is not returnable. View return policy
Catalog No. 50-811-564 Supplier New England Biolabs, Inc. Supplier No. B9003S
S-adenosyl-methionine (SAM) is included with all of the DNA methylases available from New England Biolabs, Inc. as well as certain restriction enzymes for which it is required. The SAM is prepared in a solution of 0.005 M H2SO4 and 10% ETOH and filtered.