Learn More
Lonza Walkersville S-ALI Small Airway Air-Liquid Interface Medium BulletKit
Catalog No. NC1771366
Encompass
S-ALI BulletKit Small Airway Air-Liquid Interface BulletKit provides scientists with a unique tool to investigate new areas of airway epithelial research in a more physiologically relevant way.Content:1 x S-ALI Growth Basal Media (CC-3281), 250 mL1 x S-ALI Differentiation Media (CC-3282), 500 mL1 x S-ALI SingleQuots Supplement Pack (CC-4538) containing:1 x Natural Cap Vial with Transferrin, 0.75 mL1 x Amber Vial with S-ALI-Inducer, 1.00 mL1 x Orange Cap Vial with BPE, 3.00 mL1 x Amber Vial with Epinephrine, 0.75 mL1 x Red Cap Vial with GA-1000, 0.75 mL1 x Green Cap Vial hEGF, 0.75 mL1 x Natural Cap Vial with Hydrocortisone, 0.75 mL1 x Lilac Cap Vial with Insulin, 0.75 mL1 x Amber Vial with Retinoic Acid, 0.75 mL1 x Bottle Fatty Acid Free BSA-FAF, 7.50 mL1 x Amber Vial with Triiodothyronine, 0.75 mL
Catalog No. NC1771366 Supplier Lonza Walkersville Supplier No. CC4539
Add to Cart
Add to Cart
S-ALI BulletKit Small Airway Air-Liquid Interface BulletKit provides scientists with a unique tool to investigate new areas of airway epithelial research in a more physiologically relevant way.Content:1 x S-ALI Growth Basal Media (CC-3281), 250 mL1 x S-ALI Differentiation Media (CC-3282), 500 mL1 x S-ALI SingleQuots Supplement Pack (CC-4538) containing:1 x Natural Cap Vial with Transferrin, 0.75 mL1 x Amber Vial with S-ALI-Inducer, 1.00 mL1 x Orange Cap Vial with BPE, 3.00 mL1 x Amber Vial with Epinephrine, 0.75 mL1 x Red Cap Vial with GA-1000, 0.75 mL1 x Green Cap Vial hEGF, 0.75 mL1 x Natural Cap Vial with Hydrocortisone, 0.75 mL1 x Lilac Cap Vial with Insulin, 0.75 mL1 x Amber Vial with Retinoic Acid, 0.75 mL1 x Bottle Fatty Acid Free BSA-FAF, 7.50 mL1 x Amber Vial with Triiodothyronine, 0.75 mL