Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-Allyl-L-cysteine 98.0+%, TCI America™
SDP

Catalog No. A14685G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A14685G Supplier TCI America Supplier No. A14685G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity S-Allyl-L-cysteine intended for use as a laboratory reagent.

Chemical Identifiers

CAS 21593-77-1
Molecular Formula C6H11NO2S
Molecular Weight (g/mol) 161.219
MDL Number MFCD00151975
InChI Key ZFAHNWWNDFHPOH-YFKPBYRVSA-N
Synonym (R)-Allylthio-2-aminopropionic Acid
PubChem CID 9793905
ChEBI CHEBI:74077
IUPAC Name (2R)-2-amino-3-prop-2-enylsulfanylpropanoic acid
SMILES C=CCSCC(C(=O)O)N

Specifications

Color White
Quantity 5 g
Formula Weight 161.22
Percent Purity ≥98.0% (T)
Grade Reagent
Physical Form Crystalline Powder
Chemical Name or Material S-Allyl-L-cysteine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.