Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-Alpha,Alpha-Bis[3,5-Bis(Trifluoromethyl)Phenyl]-2-Pyrrolidinemethanol Trimethylsilyl Ether

Catalog No. 6770191G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-701-91G 1 g
67-701-95G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 67-701-91G Supplier Sigma Aldrich Supplier No. 6770191G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-Alpha,Alpha-Bis[3,5-Bis(Trifluoromethyl)Phenyl]-2-Pyrrolidinemethanol Trimethylsilyl Ether

Specifications

CAS 848821-61-4
Melting Point 48°C to 56°C
Molecular Formula C24H23F12NOSi
Linear Formula C24H23F12NOSi
MDL Number MFCD09750447
Quantity 1 g
Synonym (S)-alpha,alpha-[3,5-Bis(trifluoromethyl)phenyl]prolinol trimethylsilyl ether; (S)-2-[(Bis(3,5-bis(trifluoromethyl)phenyl)trimethylsilyloxy)methyl]pyrrolidine; (S)-2-[(Bis(3,5-bis(trifluoromethyl)phenyl)trimethylsilanyloxy)methyl]pyrrolidine
Molecular Weight (g/mol) 597.51 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.