Learn More
Medchemexpress LLC (S)-Alpha-amino-2',4'-dichloro-4-hydroxy-5-(phosphonomethyl)-[1,1'-biphenyl]-3-propanoic acid | 174575-40-7 | MFCD02262130 | 99.1% | 420.18 g/mol | C16H16Cl2NO6P | 1 MG
SDP
Catalog No. 5000242099
Encompass_Preferred
SDZ 220-040 is a selective competitive antagonist of mammalian NMDA receptors (reported pKi ≈ 8.5). It is used in preclinical research to probe NMDA-mediated neurotransmission and in models of neurological disease. Typical supplier specifications report high analytical purity and small research-scale quantities for controlled laboratory use.
  • Selective competitive NMDA receptor antagonist.
  • Reported binding affinity pKi ≈ 8.5.
  • Suitable for studies of NMDA-mediated neurotransmission and neurological disease models.
  • High analytical purity for biochemical and pharmacology assays.
  • CAS number 174575-40-7 and chemical formula C16H16Cl2NO6P.
  • Intended for research use only; handle according to safety data sheets.
Catalog No. 50-002-42099 Supplier Medchemexpress LLC Supplier No. HY1077041MG
SDZ 220-040 is a selective competitive antagonist of mammalian NMDA receptors (reported pKi ≈ 8.5). It is used in preclinical research to probe NMDA-mediated neurotransmission and in models of neurological disease. Typical supplier specifications report high analytical purity and small research-scale quantities for controlled laboratory use.
  • Selective competitive NMDA receptor antagonist.
  • Reported binding affinity pKi ≈ 8.5.
  • Suitable for studies of NMDA-mediated neurotransmission and neurological disease models.
  • High analytical purity for biochemical and pharmacology assays.
  • CAS number 174575-40-7 and chemical formula C16H16Cl2NO6P.
  • Intended for research use only; handle according to safety data sheets.