Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific (S)-(-)-alpha-Methoxy-alpha-trifluoromethyl-phenylacetic acid, 17257-71-5, MFCD00064200, 1g

Catalog No. 500120191
Encompass_Preferred

Molecular Formula C10H9F3O3, Purity 99%, Molecular Weight 234.18, Melting Point 43-45°, Boiling Point 90/0.1mm

Catalog No. 50-012-0191 Supplier Matrix Scientific Supplier No. 0071681G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molecular Formula C10H9F3O3, Purity 99%, Molecular Weight 234.18, Melting Point 43-45°, Boiling Point 90/0.1mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.