Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(-)-Alpha-Methoxy-Alpha-(Trifluoromethyl)Phenylacetic Acid

Catalog No. 318035250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-803-5250MG 250 mg
31-803-51G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-803-5250MG Supplier Sigma Aldrich Supplier No. 318035250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(-)-Alpha-Methoxy-Alpha-(Trifluoromethyl)Phenylacetic Acid

Specifications

CAS 17257-71-5
Melting Point 46°C to 49°C
Density 1.303 g/mL at 25°C
Boiling Point 95°C to 97°C (0.05 mmHg, lit.)
Molecular Formula C10H9F3O3
Linear Formula C6H5C(OCH3)(CF3)CO2H
Refractive Index n20/D 1.474
MDL Number MFCD00064200
Quantity 250 mg
Synonym (-)-MTPA; Mosher's acid
Molecular Weight (g/mol) 234.17 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.