Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(+)-Alpha-Methoxy-Alpha-(Trifluoromethyl)Phenylacetyl Chloride

Catalog No. 36967550MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-967-550MG 50 mg
36-967-5250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 36-967-550MG Supplier Sigma Aldrich Supplier No. 36967550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(+)-+a-Methoxy-+a-trifluoromethylphenylacetyl chloride is commonly used as a derivatizing agent in Mosher ester analysis and Mosher amide analysis, which are NMR-based methods for determining the absolute configuration of the chiral carbon center in secondary alcohols and amines, respectively.Used in the synthesis of natural products and pheromones.doi:10.1038/nprot.2007.354

Specifications

CAS 20445-33-4
Density 1.35 g/mL at 25°C
Boiling Point 213°C to 214°C
Molecular Formula C10H8ClF3O2
Refractive Index n20/D 1.469
Linear Formula C6H5C(OCH3)(CF3)COCl
MDL Number MFCD00067105
Quantity 50 mg
Synonym (S)-(+)-MTPA-Cl; Mosher's acid chloride
Molecular Weight (g/mol) 252.62 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.