Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-alpha-Methyl-4-nitrobenzylamine Hydrochloride 99.0+%, TCI America™
SDP

Catalog No. N05431G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N05431G Supplier TCI America Supplier No. N05431G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 132873-57-5
Molecular Formula C8H11ClN2O2
Molecular Weight (g/mol) 202.64
MDL Number MFCD00066312
InChI Key CZQQGVFHLSBEDV-UHFFFAOYNA-N
Synonym (S)-Nitresolve, (S)-1-(4-Nitrophenyl)ethylamine Hydrochloride
PubChem CID 16212176
IUPAC Name 1-(4-nitrophenyl)ethan-1-amine hydrochloride
SMILES Cl.CC(N)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Quantity 1 g
Formula Weight 202.64
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-alpha-Methyl-4-nitrobenzylamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.