Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(-)-Alpha-Methylbenzyl Isocyanate

Catalog No. 2205661G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-056-61G 1 g
22-056-65G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 22-056-61G Supplier Sigma Aldrich Supplier No. 2205661G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(-)-+a-Methylbenzyl isocyanate is a commonly used chiral derivatizing agent for the resolution of enantiomers.(S)-(-)-+a-Methylbenzyl isocyanate may be used as a chiral auxiliary for the resolution of racemic (+/-)-6aR,11aR-maackiain to give the (-)-form, which is a key intermediate for synthesizing (-)-cabenegrin A-I.

Specifications

CAS 14649-03-7
Density 1.045 g/mL at 20°C
Boiling Point 55°C to 56°C (2.5 mmHg, lit.)
Molecular Formula C9H9NO
Refractive Index n20/D 1.5145
Linear Formula C6H5CH(CH3)NCO
MDL Number MFCD00002036
Quantity 1 g
Synonym (S)-(-)-1-Phenylethyl isocyanate
Molecular Weight (g/mol) 147.17 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.