Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(+)-Alpha-(Trifluoromethyl)Benzyl Alcohol

Catalog No. 4111401G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-114-01G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 41-114-01G Supplier Sigma Aldrich Supplier No. 4111401G
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(+)-Alpha-(Trifluoromethyl)Benzyl Alcohol

Specifications

CAS 1433-27-8
Density 0.814 g/mL at 25°C
Boiling Point 137°C to 139°C
Molecular Formula C7H14N2
Refractive Index n20/D 1.462
Linear Formula (CH3)3CN=C=NC2H5
MDL Number MFCD00209556
Quantity 1 g
Synonym (+)-Phenyl(trifluoromethyl)carbinol; (S)-(+)-1-Phenyl-2,2,2-trifluoroethanol
Molecular Weight (g/mol) 176.14 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.