Learn More
Medchemexpress LLC (βS)-β-amino-4-[(3,4-dichlorophenyl)methoxy]-β-methyl-benzenebutanol | 2748196-63-4 | MFCD00006370 | 97.0% | 354.27 g·mol^-1 | C18H21Cl2NO2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY12601510MG
Encompass_Preferred
P053 is a selective, non-competitive inhibitor of ceramide synthase 1 (CerS1) used in biochemical and cellular research to probe ceramide metabolism, mitochondrial fatty acid oxidation, and adiposity regulation. Reported IC50 is approximately 0.5 μM. The compound is supplied as a white to off-white solid at high purity for in vitro research use.
Catalog No. 50-002-38235