Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-β-Aminoisobutyric acid (hydrochloride) | 925704-45-6 | MFCD12198195 | 95.0% | 139.58 | C4H10ClNO2 | 250 MG
SDP

Catalog No. 5000302379
Encompass_Preferred

(S)-β-Aminoisobutyric acid hydrochloride is the S-enantiomer of β-aminoisobutyric acid, a non-proteinogenic amino acid produced during thymine and valine catabolism. Provided as the hydrochloride salt for research use, it serves as an endogenous metabolite standard and a tool in studies of metabolic enzymes and protease pathways.

  • High purity suitable for research applications.
  • Hydrochloride salt form improves solubility and stability.
  • Molecular formula C4H10ClNO2; molecular weight 139.58.
  • Storage: room temperature up to 3 years; in solvent, -80°C for 2 years or -20°C for 1 year.
  • Intended for biochemical and metabolic pathway research.

Catalog No. 50-003-02379 Supplier Medchemexpress LLC Supplier No. 250MGHYW023323
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-β-Aminoisobutyric acid hydrochloride is the S-enantiomer of β-aminoisobutyric acid, a non-proteinogenic amino acid produced during thymine and valine catabolism. Provided as the hydrochloride salt for research use, it serves as an endogenous metabolite standard and a tool in studies of metabolic enzymes and protease pathways.

  • High purity suitable for research applications.
  • Hydrochloride salt form improves solubility and stability.
  • Molecular formula C4H10ClNO2; molecular weight 139.58.
  • Storage: room temperature up to 3 years; in solvent, -80°C for 2 years or -20°C for 1 year.
  • Intended for biochemical and metabolic pathway research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.