Learn More
Biomatik Corporation S Recombinant Antibody
Supplier:  Biomatik Corporation RAC07047
Encompass_Preferred
S Recombinant Antibody is a Human (Homo sapiens) Novel Coronavirus (SARS-CoV-2/ 2019-nCoV) Recombinant Antibody to S. The S Recombinant Antibody has been validated for the following applications: ELISA, GICA, Neutralising. Host: HEK293F Cell. Species: Human (Homo sapiens) Novel Coronavirus (SARS-CoV-2/ 2019-nCoV). Immunogen: Recombinant Human Novel Coronavirus Spike Glycoprotein (S) (16-685Aa) (Csb-Mp3324Gmy). Applications: ELISA, GICA, Neutralising. Target Synonyms: S. Accession Number: P0DTC2. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9267