Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-Antphos

Catalog No. 912476100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-247-6100MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-247-6100MG Supplier Sigma Aldrich Supplier No. 912476100MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-AntPhos is a P-chiral monophosphorus ligand used for the asymmetric Suzuki-Miyaura and Miyaura borylation reactions. This ligand is uniquely effective for sterically hindered cross-coupling reactions.Synthesis of Chiral alpha-Amino Tertiary Boronic Esters by Enantioselective Hydroboration of alpha-ArylenamidesSold in collaboration with Zejun Pharmaceuticals

Specifications

CAS 228758-57-3
Molecular Formula C19H24N4O4 -+ xHCl
Linear Formula C24H32O4P2
Quantity 50 mg
Synonym 4-((6-Aminohexyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride; Crosslinker-E3 ligase ligand conjugate; Pomalidomide conjugate; Protein degrader building block for PROTAC(R) research; Template for synthesis of targeted protein degrader
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.