Learn More
INTACT GENOMICS INC Staphylococcus aureus (S. aureus) RN4220 Electrocompetent Cells 5x 200µl
Catalog No. 5000082950
Encompass
Intact Genomics (ig) Staphylococcus aureus (S. aureus) RN4220 Electrocompetent Cells are optimized to provide high transformation efficiencies making them ideal for various applications. This strain is commonly used in studies involving virulence, resistance, metabolism and more. Staphylococcus aureus RN4220 is characterized by a mutation in the sau1 hsdR genes, making it restriction deficient and hence an excellent intermediate cloning host.Product Include and Storage:ig Staphylococcus aureus (S. aureus) RN4220 Electrocompetent CellsStaph Recovery mediumBSL2 facility is required for purchase and use of this product.
Catalog No. 50-000-82950 Supplier INTACT GENOMICS INC Supplier No. 129440
Intact Genomics (ig) Staphylococcus aureus (S. aureus) RN4220 Electrocompetent Cells are optimized to provide high transformation efficiencies making them ideal for various applications. This strain is commonly used in studies involving virulence, resistance, metabolism and more. Staphylococcus aureus RN4220 is characterized by a mutation in the sau1 hsdR genes, making it restriction deficient and hence an excellent intermediate cloning host.Product Include and Storage:ig Staphylococcus aureus (S. aureus) RN4220 Electrocompetent CellsStaph Recovery mediumBSL2 facility is required for purchase and use of this product.