Learn More
AdipoGen S-Benzoylthiamine O-PO4
SDP
Catalog No. 502561596
Encompass_Preferred
Chemical. CAS 22457-89-2. Formula C19H23N4O6PS. MW 466.45. S-Benzoylthiamine O-monophosphate is a lipid-soluble form of vitamin B1 thiamine. S-Benzoylthiamine O-monophosphate facilitates the action of thiamine diphosphate, a cofactor for the enzyme transketolase. The activation of transketolase enzyme accelerates the precursors of advanced glycation end products AGEs towards the pentose phosphate pathway thereby reducing the production of AGEs. The reduction in AGEs subsequently decreases metabolic stress which benefits vascular complications seen in diabetes. The anti-AGE property of S-Benzoylthiamine O-monophosphate certainly makes it effective for the treatment of diabetic neuropathy, nephropathy and retinopathy. S-Benzoylthiamine O-monophosphate appears to have a also cardioprotective role in potentiating angiogenesis and inhibiting apoptosis in diabetic complications.
Catalog No. 50-256-1596 Supplier AdipoGen Supplier No. CDXB0403G005
Chemical. CAS 22457-89-2. Formula C19H23N4O6PS. MW 466.45. S-Benzoylthiamine O-monophosphate is a lipid-soluble form of vitamin B1 thiamine. S-Benzoylthiamine O-monophosphate facilitates the action of thiamine diphosphate, a cofactor for the enzyme transketolase. The activation of transketolase enzyme accelerates the precursors of advanced glycation end products AGEs towards the pentose phosphate pathway thereby reducing the production of AGEs. The reduction in AGEs subsequently decreases metabolic stress which benefits vascular complications seen in diabetes. The anti-AGE property of S-Benzoylthiamine O-monophosphate certainly makes it effective for the treatment of diabetic neuropathy, nephropathy and retinopathy. S-Benzoylthiamine O-monophosphate appears to have a also cardioprotective role in potentiating angiogenesis and inhibiting apoptosis in diabetic complications.