Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-Benzylisothiouronium chloride, 98+%, Thermo Scientific™

Catalog No. AAA1591418
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1591418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1591418 Supplier Thermo Scientific Chemicals Supplier No. A1591418

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It has been studied as a small-molecule inhibitor of indoleamine-2,3-dioxygenase.

Solubility
Soluble in water and methanol.

Notes
Hygroscopic. Store in cool dry place. Keep away from oxidizing agent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 538-28-3
Molecular Formula C8H11ClN2S
Molecular Weight (g/mol) 202.7
MDL Number MFCD00012583
InChI Key WJAASTDRAAMYNK-UHFFFAOYSA-N
Synonym s-benzylisothiourea hydrochloride, 2-benzyl-2-thiopseudourea hydrochloride, btkh, s-benzylthiuronium chloride, benzyl carbamimidothioate hydrochloride, s-benzylthiouronium chloride, benzylisothiourea hydrochloride, benzylthiuronium chloride, 2-benzylisothiouronium chloride, benzylisothiuronium chloride
PubChem CID 10852
IUPAC Name benzyl carbamimidothioate;hydrochloride
SMILES C1=CC=C(C=C1)CSC(=N)N.Cl

Specifications

Melting Point 173°C to 176°C
Quantity 50 g
UN Number UN2811
Beilstein 3656719
Merck Index 14,1146
Formula Weight 202.7
Percent Purity ≥98%
Chemical Name or Material S-Benzylisothiouronium chloride
Hazard Category Danger
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECSNumber UM0738000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.