Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[(S)-Binap RuCl benzene]Cl, Thermo Scientific™

Catalog No. AAH3560903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3560903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3560903 Supplier Thermo Scientific Chemicals Supplier No. H3560903

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126251-92-1
Molecular Formula C50H38Cl2P2Ru
Molecular Weight (g/mol) 872.772
MDL Number MFCD22419153
InChI Key ZXBWEYDPYJZEFT-UHFFFAOYSA-L
Synonym s-binap rucl benzene cl, c50h38p2rucl2, s-rucl benzene binap cl, benzene binap chlororutheniumylium chloride, dichloro benzene s-2,2'-bis diphenylphosphino-1,1'-binaphthyl ruthenium ii, benzene;chlororuthenium 1+ ; 1-2-diphenylphosphanylnaphthalen-1-yl naphthalen-2-yl-diphenylphosphane;chloride
PubChem CID 11072956
IUPAC Name benzene;chlororuthenium(1+);[1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane;chloride
SMILES C1=CC=CC=C1.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.[Cl-].Cl[Ru+]

Specifications

Quantity 1 g
Formula Weight 872.8
Chemical Name or Material [(S)-Binap RuCl benzene]Cl
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 664-455-2
TSCA No
Recommended Storage Ambient temperatures; Store under Argon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.